Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BQ024807.1 |
- |
1 |
|
|
Chr. 15 - 34130372 - 34131066 [+] on Build GRCh38 |
99 |
GenBankmRNA |
AK091619.1 |
- |
|
2143 |
|
Chr. - - [-] on Build |
99 |
GenBankmRNA |
CA949694.1 |
- |
1 |
370 |
|
Chr. 15 - 34130773 - 34131215 [-] on Build GRCh38 |
99 |
GenBankmRNA |
DB302990.1 |
- |
1 |
|
|
Chr. 15 - 34130473 - 34131045 [+] on Build GRCh38 |
99 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
piggyBac transposable element derived 4 |
PGBD4 |
|
-26.1kb
|
Sense |
Chr. - 34102073 - 34104390 on Build GRCh38 |
Downstream |
katanin regulatory subunit B1 like 1 |
KATNBL1 |
C15orf29 |
9.6kb
|
Anti-sense |
Chr. - 34140674 - 34210146 on Build GRCh38 |
View Full Product Details