Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZBTB20
Entrez Gene ID
26137
Gene Name
zinc finger and BTB domain containing 20
Gene Aliases
DPZF, HOF, ODA-8S, PRIMS, ZNF288
Chromosome Location
Chr.3: 114314500 - 115147280 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164342.2 NP_001157814.1 5 - 5 26766 118
NM_001164343.2 NP_001157815.1 12 - 12 27228 118
NM_001164344.2 NP_001157816.1 8 - 8 27192 118
NM_001164345.2 NP_001157817.1 7 - 7 27146 118
NM_001164346.2 NP_001157818.1 6 - 6 26785 118
NM_001164347.2 NP_001157819.1 6 - 6 26807 118
NM_015642.5 NP_056457.3 10 - 10 26990 118
NR_121662.1 7 - 7 25248 118
GenBank mRNA AL049423.1 - 1 - 1 1544 118


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AI128131.1 - 1 Chr. 3 - 114314501 - 114314969 [+] on Build GRCh38 118
GenBankmRNA H03919.1 - 1 Chr. 3 - 114314519 - 114314886 [+] on Build GRCh38 118
GenBankmRNA AI089579.1 - 1 Chr. 3 - 114314501 - 114314884 [+] on Build GRCh38 118
GenBankmRNA DB306784.1 - 1 Chr. 3 - 114314506 - 114315029 [+] on Build GRCh38 118
GenBankmRNA CD678022.1 - 1 467 Chr. 3 - 114314500 - 114315102 [-] on Build GRCh38 118
GenBankmRNA H04645.1 - 1 Chr. 3 - 114314552 - 114314861 [+] on Build GRCh38 118
GenBankmRNA AA015653.1 - 1 Chr. 3 - 114314501 - 114314787 [+] on Build GRCh38 119
GenBankmRNA AA281591.1 - 1 Chr. 3 - 114314512 - 114314876 [+] on Build GRCh38 118
GenBankmRNA AW003707.1 - 1 Chr. 3 - 114314500 - 114315009 [+] on Build GRCh38 118
GenBankmRNA BI492767.1 - 1 Chr. 3 - 114314501 - 114314769 [+] on Build GRCh38 118
GenBankmRNA BQ026125.1 - 1 Chr. 3 - 114314500 - 114314944 [+] on Build GRCh38 117
GenBankmRNA CA774229.1 - 1 Chr. 3 - 114314501 - 114314964 [+] on Build GRCh38 118
GenBankmRNA AI192694.1 - 1 Chr. 3 - 114314501 - 114314856 [+] on Build GRCh38 118
GenBankmRNA HY315389.1 - 1 Chr. 3 - 114314548 - 114314983 [+] on Build GRCh38 118
GenBankmRNA H20263.1 - 1 Chr. 3 - 114314515 - 114314864 [+] on Build GRCh38 118
GenBankmRNA AI282344.1 - 1 Chr. 3 - 114314502 - 114314799 [+] on Build GRCh38 118
GenBankmRNA AA766016.1 - 1 Chr. 3 - 114314513 - 114314853 [+] on Build GRCh38 118
GenBankmRNA C16569.1 - 128 Chr. 11 - 114441889 - 114441996 [+] on Build GRCh38 118
GenBankmRNA AW021837.1 - 1 134 Chr. 3 - 114314501 - 114314769 [-] on Build GRCh38 118


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream T-cell immunoreceptor with Ig and ITIM domains TIGIT VSIG9, VSTM3, WUCAM -4.2kb Anti-sense Chr. - 114293986 - 114310288 on Build GRCh38
Downstream microRNA 568 MIR568 MIRN568, hsa-mir-568 296.0 Sense Chr. - 114316475 - 114316569 on Build GRCh38

View Full Product Details