Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AL713779.1 - 1 1947 Chr. 3 - 128550049 - 128552359 [+] on Build GRCh38 103


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream uncharacterized LOC90246 LOC90246 -39.5kb Sense Chr. - 128507835 - 128510595 on Build GRCh38
Downstream long intergenic non-protein coding RNA 1565 LINC01565 C3orf27, GR6 19.6kb Anti-sense Chr. - 128572000 - 128576086 on Build GRCh38

View Full Product Details