The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_110428.1 | 3 - 3 | 397 | 87 | |
NR_110429.1 | 3 - 3 | 423 | 87 | ||
GenBank mRNA | AJ707365.1 | - | 3 - 3 | 360 | 87 |
BM782281.1 | - | 3 - 3 | 397 | 87 | |
BX107736.1 | - | 3 - 3 | 335 | 87 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | TGFB induced factor homeobox 1 | TGIF1 | HPE4, TGIF | -20.3kb | Anti-sense | Chr. - 3411927 - 3458412 on Build GRCh38 |
Downstream | DLG associated protein 1 | DLGAP1 | DAP-1, DAP-1-ALPHA, DAP-1-BETA, DAP1, DLGAP1A, DLGAP1B, GKAP, SAPAP1 | 17.1kb | Sense | Chr. - 3496032 - 4455307 on Build GRCh38 |