Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BG547809.1 - 1 543 Chr. 9 - 4258368 - 4259207 [-] on Build GRCh38 100
GenBankmRNA BC017428.1 - 1 547 Chr. 9 - 4258106 - 4259207 [-] on Build GRCh38 101


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream GLIS family zinc finger 3 GLIS3 NDH, ZNF515 Sense Chr. - 3824127 - 4310694 on Build GRCh38
Downstream solute carrier family 1 member 1 SLC1A1 DCBXA, EAAC1, EAAT3, SCZD18 231.2kb Anti-sense Chr. - 4490427 - 4587469 on Build GRCh38

View Full Product Details