Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA HX785781.1 - 1 349 Chr. 21 - 14545590 - 14546312 [-] on Build GRCh38 105
GenBankmRNA BG530126.1 - 1 352 Chr. 21 - 14545573 - 14546315 [-] on Build GRCh38 105
GenBankmRNA CF551990.1 - 1 340 Chr. 21 - 14545441 - 14546277 [-] on Build GRCh38 105
GenBankmRNA AV733685.1 - 1 239 Chr. 21 - 14545563 - 14546203 [-] on Build GRCh38 105
GenBankmRNA HX789251.1 - 1 349 Chr. 21 - 14545590 - 14546312 [-] on Build GRCh38 105
GenBankmRNA BC020638.1 - 1 352 Chr. 21 - 14545145 - 14546315 [-] on Build GRCh38 105


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream SAM domain, SH3 domain and nuclear localization signals 1 SAMSN1 HACS1, NASH1, SASH2, SH3D6B, SLy2 Sense Chr. - 14485228 - 14583402 on Build GRCh38
Downstream SAMSN1 antisense RNA 1 SAMSN1-AS1 35.9kb Anti-sense Chr. - 14582202 - 14598303 on Build GRCh38

View Full Product Details