Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- SRRM2
- Entrez Gene ID
-
23524
- Gene Name
- serine/arginine repetitive matrix 2
- Gene Aliases
-
300-KD, CWF21, Cwc21, HSPC075, SRL300, SRm300
- Chromosome Location
-
Chr.16: 2752329 - 2771412 on Build GRCh38
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BE671072.1 |
- |
1 |
|
|
Chr. 16 - 2759568 - 2760845 [-] on Build GRCh38 |
89 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
SRRM2 antisense RNA 1 |
SRRM2-AS1 |
|
-7.3kb
|
Anti-sense |
Chr. - 2737076 - 2752600 on Build GRCh38 |
Downstream |
transcription elongation factor B subunit 2 |
TCEB2 |
ELOB, SIII |
10.6kb
|
Anti-sense |
Chr. - 2771414 - 2777296 on Build GRCh38 |
View Full Product Details