Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA Y12839.1 - 1 3552 Chr. 2 - 74578858 - 74582566 [-] on Build GRCh38 122


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream meiosis 1 associated protein M1AP C2orf65, D6Mm5e, SPATA37 Sense Chr. - 74557640 - 74648338 on Build GRCh38
Downstream ssemaphorin 4F SEMA4F M-SEMA, PRO2353, S4F, SEMAM, SEMAW, m-Sema-M 71.7kb Anti-sense Chr. - 74654228 - 74699013 on Build GRCh38

View Full Product Details