Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- RNA18S5
- Entrez Gene ID
-
100008588
- Gene Name
- RNA, 18S ribosomal 5
- Gene Aliases
-
RN18S1
- Chromosome Location
-
Chr.Un|NT_167214: 109078 - 110946 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_003286.2 |
|
1 - 1 |
46 |
122 |
- Gene Symbol
- RNA45S5
- Entrez Gene ID
-
100861532
- Gene Name
- RNA, 45S pre-ribosomal 5
- Gene Aliases
-
RN45S
- Chromosome Location
-
Chr.Un|NT_167214: 105424 - 118780 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_046235.1 |
|
1 - 1 |
3700 |
122 |
View Full Product Details