The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022469.3 | NP_071914.3 | 2 - 2 | 1242 | 96 |
XM_005273226.3 | XP_005273283.1 | 2 - 2 | 1070 | 96 | |
XM_011544249.2 | XP_011542551.1 | 3 - 3 | 1572 | 96 | |
GenBank mRNA | AK024848.1 | - | 2 - 2 | 1229 | 96 |
BC046632.1 | - | 2 - 2 | 1169 | 96 |