The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000479.3 | NP_000470.2 | 1 - 1 | 329 | 103 |
GenBank mRNA | AI791949.1 | - | 1 - 1 | 529 | 102 |
BC049194.1 | - | 1 - 1 | 132 | 103 |