The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001085486.1 | NP_001078955.1 | 3 - 3 | 154 | 177 |
NM_001093726.1 | NP_001087195.1 | 3 - 3 | 213 | 177 | |
NM_005410.2 | NP_005401.3 | 2 - 2 | 125 | 177 | |
GenBank mRNA | AK096125.1 | - | 2 - 2 | 494 | 177 |
AK225801.1 | - | 3 - 3 | 124 | 177 | |
AK311392.1 | - | 2 - 2 | 462 | 177 | |
AL833145.1 | - | 2 - 2 | 96 | 177 | |
AU136519.2 | - | 3 - 3 | 183 | 177 | |
BC005244.1 | - | 2 - 2 | 95 | 177 | |
BC015875.1 | - | 2 - 2 | 99 | 177 | |
BC040075.1 | - | 2 - 2 | 75 | 177 | |
BC046152.1 | - | 2 - 2 | 76 | 177 | |
BC058919.1 | - | 2 - 2 | 106 | 177 | |
BI463468.1 | - | 2 - 2 | 270 | 177 | |
Z11793.1 | - | 2 - 2 | 61 | 177 |