The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001165967.1 | NP_001159439.1 | 2 - 3 | 148 | 90 |
NM_032580.3 | NP_115969.2 | 2 - 3 | 148 | 90 | |
XM_011524041.2 | XP_011522343.1 | 3 - 4 | 686 | 90 | |
XM_017025232.1 | XP_016880721.1 | 3 - 4 | 1742 | 90 | |
GenBank mRNA | AB049064.1 | - | 2 - 3 | 140 | 90 |
BC039887.1 | - | 2 - 3 | 146 | 90 | |
BI259590.1 | - | 3 - 4 | 334 | 90 |