The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166243.2 | NP_001159715.1 | 9 - 10 | 822 | 70 |
XM_017005884.1 | XP_016861373.1 | 10 - 11 | 870 | 70 | |
GenBank mRNA | BC032336.1 | - | 9 - 10 | 774 | 70 |