Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UPK2
Entrez Gene ID
7379
Gene Name
uroplakin 2
Gene Aliases
UP2, UPII
Chromosome Location
Chr.11: 118956298 - 118958559 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006760.3 NP_006751.1 2 - 3 260 58
GenBank mRNA AB009454.1 - 2 - 3 147 58
AY455312.1 - 2 - 3 207 58
BC113900.1 - 2 - 3 238 58
CD556028.1 - 2 - 3 275 58
Y13645.1 - 2 - 3 245 58

View Full Product Details