The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021571.3 | NP_067546.1 | 1 - 2 | 279 | 66 |
GenBank mRNA | AI189838.1 | - | 1 - 2 | 66 | |
AW295205.1 | - | 1 - 2 | 66 | ||
AY358231.1 | - | 1 - 2 | 283 | 66 | |
BC131610.1 | - | 1 - 2 | 271 | 66 |