The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022117.3 | NP_071400.1 | 4 - 5 | 1276 | 87 |
XM_006724592.3 | XP_006724655.1 | 4 - 5 | 1309 | 87 | |
XM_017029726.1 | XP_016885215.1 | 3 - 4 | 1441 | 87 | |
XM_017029727.1 | XP_016885216.1 | 3 - 4 | 1441 | 87 | |
GenBank mRNA | AB015345.1 | - | 4 - 5 | 516 | 87 |
AF254794.1 | - | 4 - 5 | 1015 | 87 | |
AF273046.1 | - | 4 - 5 | 1266 | 87 | |
AK131220.1 | - | 3 - 4 | 1414 | 87 | |
AK292462.1 | - | 4 - 5 | 1269 | 87 | |
AL713652.1 | - | 4 - 5 | 1255 | 87 | |
AY040871.1 | - | 4 - 5 | 1265 | 87 | |
BC001566.1 | - | 1 - 2 | 73 | 87 | |
BC024270.1 | - | 4 - 5 | 1256 | 87 |