The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173588.3 | NP_775859.3 | 15 - 16 | 2595 | 83 |
XM_011520030.2 | XP_011518332.1 | 13 - 14 | 5499 | 83 | |
XM_017017607.1 | XP_016873096.1 | 14 - 15 | 5390 | 83 | |
XM_017017608.1 | XP_016873097.1 | 14 - 15 | 3413 | 83 | |
GenBank mRNA | BC115396.1 | - | 15 - 16 | 2546 | 83 |
BC115397.1 | - | 15 - 16 | 2546 | 83 |