The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173588.3 | NP_775859.3 | 6 - 7 | 709 | 92 |
XM_011520030.2 | XP_011518332.1 | 4 - 5 | 3613 | 92 | |
XM_011520031.2 | XP_011518333.1 | 4 - 5 | 3613 | 92 | |
XM_017017607.1 | XP_016873096.1 | 5 - 6 | 3504 | 92 | |
XM_017017608.1 | XP_016873097.1 | 5 - 6 | 1527 | 92 | |
GenBank mRNA | AK095113.1 | - | 6 - 7 | 709 | 92 |
BC115396.1 | - | 6 - 7 | 660 | 92 | |
BC115397.1 | - | 6 - 7 | 660 | 92 |