The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_176812.4 | NP_789782.1 | 3 - 4 | 645 | 93 |
GenBank mRNA | AB100261.1 | - | 3 - 4 | 590 | 93 |
AY329085.1 | - | 3 - 4 | 480 | 93 | |
BC033859.1 | - | 3 - 4 | 589 | 93 |