Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IQCF5
Entrez Gene ID
389124
Gene Name
IQ motif containing F5
Gene Aliases
-
Chromosome Location
Chr.3: 51873721 - 51875584 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145059.1 NP_001138531.1 2 - 2 451 88
GenBank mRNA BX106781.1 - 2 - 2 451 88

View Full Product Details