The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002563.4 | NP_002554.1 | 1 - 1 | 1366 | 115 |
GenBank mRNA | AK313920.1 | - | 1 - 1 | 551 | 115 |
AY136752.1 | - | 1 - 1 | 276 | 115 | |
BC074784.2 | - | 1 - 1 | 298 | 115 | |
BC074785.2 | - | 1 - 1 | 298 | 115 | |
U42029.1 | - | 1 - 1 | 322 | 115 | |
U42030.1 | - | 1 - 1 | 413 | 115 | |
Z49205.1 | - | 1 - 1 | 1125 | 115 |