Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
P2RY4
Entrez Gene ID
5030
Gene Name
pyrimidinergic receptor P2Y4
Gene Aliases
NRU, P2P, P2Y4, UNR
Chromosome Location
Chr.X: 70258166 - 70259804 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002565.3 NP_002556.1 1 - 1 488 110
GenBank mRNA BC095503.1 - 1 - 1 447 110
BC096067.3 - 1 - 1 351 110
BC096068.1 - 1 - 1 351 110
BC096069.1 - 1 - 1 351 110
BC096070.1 - 1 - 1 351 110

View Full Product Details