The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001172632.1 | NP_001166103.1 | 3 - 4 | 531 | 84 |
GenBank mRNA | AK295409.1 | - | 3 - 4 | 472 | 84 |
AK316353.1 | - | 2 - 3 | 261 | 84 |