The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166346.1 | NP_001159818.3 | 2 - 3 | 683 | 110 |
GenBank mRNA | BC040713.1 | - | 1 - 2 | 273 | 110 |
CB048531.1 | - | 2 - 3 | 275 | 110 |