Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LILRA3
Entrez Gene ID
11026
Gene Name
leukocyte immunoglobulin like receptor A3
Gene Aliases
CD85E, HM31, HM43, ILT-6, ILT6, LIR-4, LIR4
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172654.2 NP_001166125.1 7 - 7 954 96
NM_006865.4 NP_006856.3 7 - 7 1190 96
GenBank mRNA AF014923.1 - 7 - 7 1098 96
AF014924.1 - 7 - 7 906 96
AF025527.1 - 7 - 7 1191 96
BC028208.1 - 7 - 7 1163 96
U91926.1 - 7 - 7 1160 96

View Full Product Details