Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- STOML1
- Entrez Gene ID
-
9399
- Gene Name
- stomatin like 1
- Gene Aliases
-
SLP-1, STORP, hUNC-24
- Chromosome Location
-
Chr.15: 73983218 - 73994622 on Build GRCh38
View Full Product Details