Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
STOML1
Entrez Gene ID
9399
Gene Name
stomatin like 1
Gene Aliases
SLP-1, STORP, hUNC-24
Chromosome Location
Chr.15: 73983218 - 73994622 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256672.1 NP_001243601.1 3 - 3 434 101
NM_001256675.1 NP_001243604.1 3 - 3 434 101
NM_001256676.1 NP_001243605.1 3 - 3 248 101
NM_001256677.1 NP_001243606.1 4 - 4 565 101
NM_001324226.1 NP_001311155.1 3 - 3 248 101
NM_001324227.1 NP_001311156.1 3 - 3 248 101
NM_001324228.1 NP_001311157.1 3 - 3 312 101
NM_001324229.1 NP_001311158.1 3 - 3 434 101
NM_001324230.1 NP_001311159.1 4 - 4 565 101
NM_004809.4 NP_004800.2 3 - 3 434 101
XM_005254792.4 XP_005254849.1 3 - 3 1338 101
XM_006720773.3 XP_006720836.1 3 - 3 1339 101
GenBank mRNA AF074953.1 - 3 - 3 296 101
AF156557.1 - 3 - 3 442 101
AK075244.1 - 3 - 3 330 101
AK091531.1 - 3 - 3 234 101
AK222665.1 - 3 - 3 317 101
AK300799.1 - 4 - 4 449 101
AL109665.1 - 3 - 3 288 101
AL533460.3 - 3 - 3 225 101
BC034379.1 - 3 - 3 380 101
BC065249.1 - 3 - 3 380 101
BX464941.2 - 3 - 3 248 101
CR542089.1 - 3 - 3 309 101
DQ064605.1 - 3 - 3 309 101
Y16522.1 - 3 - 3 353 101

View Full Product Details