The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000956.3 | NP_000947.2 | 1 - 2 | 1089 | 92 |
GenBank mRNA | AY275471.1 | - | 1 - 2 | 838 | 92 |
BC093927.1 | - | 1 - 2 | 891 | 92 | |
BC093929.1 | - | 1 - 2 | 891 | 92 | |
U19487.1 | - | 1 - 2 | 994 | 92 | |
X83868.1 | - | 1 - 2 | 838 | 92 |