Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OR6Y1
Entrez Gene ID
391112
Gene Name
olfactory receptor family 6 subfamily Y member 1
Gene Aliases
OR1-11, OR6Y2
Chromosome Location
Chr.1: 158547128 - 158548105 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005189.1 NP_001005189.1 1 - 1 651 62

View Full Product Details