Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ANKRD34A
Entrez Gene ID
284615
Gene Name
ankyrin repeat domain 34A
Gene Aliases
ANKRD34
Chromosome Location
Chr.1: 145959441 - 145964582 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039888.3 NP_001034977.1 2 - 3 629 81
GenBank mRNA AK128050.1 - 2 - 3 629 81

View Full Product Details