The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040059.1 | NP_001035148.1 | 2 - 3 | 676 | 95 |
NM_001251.2 | NP_001242.2 | 2 - 3 | 758 | 95 | |
GenBank mRNA | AK222492.1 | - | 2 - 3 | 605 | 95 |
AK222514.1 | - | 2 - 3 | 604 | 95 | |
AK290238.1 | - | 2 - 3 | 618 | 95 | |
AK298055.1 | - | 3 - 4 | 565 | 95 | |
AK301223.1 | - | 2 - 3 | 510 | 95 | |
BC015557.2 | - | 2 - 3 | 630 | 95 | |
BT009923.1 | - | 2 - 3 | 567 | 95 | |
BX438711.2 | - | 2 - 3 | 515 | 95 | |
BX647524.1 | - | 2 - 3 | 525 | 95 | |
S57235.1 | - | 2 - 3 | 582 | 95 |