Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FAM71C
Entrez Gene ID
196472
Gene Name
family with sequence similarity 71 member C
Gene Aliases
-
Chromosome Location
Chr.12: 99647750 - 99650114 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153364.3 NP_699195.1 1 - 2 1043 77
GenBank mRNA AK312767.1 - 1 - 2 1039 77
BC031221.1 - 1 - 2 1040 77

View Full Product Details