The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001083600.2 | NP_001077069.1 | 6 - 7 | 1141 | 90 |
NM_001083601.2 | NP_001077070.1 | 7 - 8 | 1241 | 90 | |
NM_001317093.1 | NP_001304022.1 | 6 - 7 | 1115 | 90 | |
NM_001317098.1 | NP_001304027.1 | 5 - 6 | 843 | 90 | |
GenBank mRNA | AK095819.1 | - | 2 - 3 | 490 | 90 |
AK130792.1 | - | 6 - 7 | 1048 | 90 | |
AK297219.1 | - | 6 - 7 | 1044 | 90 | |
AL833594.1 | - | 7 - 8 | 1118 | 90 | |
AY358543.1 | - | 6 - 7 | 1037 | 90 | |
BC011267.1 | - | 5 - 6 | 954 | 90 | |
BG472911.1 | - | 2 - 3 | 466 | 90 | |
BG822206.1 | - | 5 - 6 | 699 | 90 |