Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TLX1NB
Entrez Gene ID
100038246
Gene Name
TLX1 neighbor
Gene Aliases
APT-B7, TD1, TDI
Chromosome Location
Chr.10: 101089321 - 101141266 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_130722.1 2 - 3 527 96
NR_130723.1 2 - 3 498 96
GenBank mRNA BI770592.1 - 2 - 3 508 96

View Full Product Details