Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLSCR5
Entrez Gene ID
389158
Gene Name
phospholipid scramblase family member 5
Gene Aliases
-
Chromosome Location
Chr.3: 146576555 - 146606216 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001085420.1 NP_001078889.1 2 - 3 1190 94
XM_017006373.1 XP_016861862.1 2 - 3 401 94

View Full Product Details