Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MRGPRF
Entrez Gene ID
116535
Gene Name
MAS related GPR family member F
Gene Aliases
GPR140, GPR168, MRGF, RTA
Chromosome Location
Chr.11: 69004375 - 69013382 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098515.1 NP_001091985.1 1 - 2 298 48
GenBank mRNA AK122669.1 - 1 - 2 172 48

View Full Product Details