Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATG101
Entrez Gene ID
60673
Gene Name
autophagy related 101
Gene Aliases
C12orf44
Chromosome Location
Chr.12: 52069974 - 52077495 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098673.1 NP_001092143.1 1 - 2 102 100
GenBank mRNA BP309508.1 - 1 - 2 73 100

View Full Product Details