The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001098673.1 | NP_001092143.1 | 3 - 4 | 554 | 61 |
NM_021934.4 | NP_068753.2 | 3 - 4 | 725 | 61 | |
GenBank mRNA | AF218031.1 | - | 3 - 4 | 666 | 61 |
AK021835.1 | - | 3 - 4 | 723 | 61 | |
BC005151.2 | - | 3 - 4 | 677 | 61 | |
BC009937.2 | - | 3 - 4 | 659 | 61 | |
BP309508.1 | - | 3 - 4 | 525 | 61 |