The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001099434.1 | NP_001092904.1 | 5 - 6 | 674 | 93 |
GenBank mRNA | BC041458.1 | - | 3 - 4 | 1192 | 93 |
BC128073.1 | - | 5 - 6 | 674 | 93 |