The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001277947.1 | NP_001264876.1 | 5 - 6 | 502 | 76 |
NM_001277951.1 | NP_001264880.1 | 2 - 3 | 213 | 76 | |
NM_018300.3 | NP_060770.3 | 2 - 3 | 162 | 76 | |
GenBank mRNA | AK027518.1 | - | 2 - 3 | 207 | 76 |
BX647540.1 | - | 1 - 2 | 2019 | 76 | |
DA701045.1 | - | 5 - 6 | 407 | 76 | |
DA731895.1 | - | 2 - 3 | 207 | 76 | |
DR761375.1 | - | 1 - 2 | 100 | 76 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_011527096.2 | XP_011525398.1 | 5 - 6 | 2422 | 76 |