Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NACA
Entrez Gene ID
4666
Gene Name
nascent polypeptide-associated complex alpha subunit
Gene Aliases
HSD48, NAC-alpha, NACA1, skNAC
Chromosome Location
Chr.12: 56712427 - 56726275 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113201.2 NP_001106672.1 1 - 2 332 71
GenBank mRNA BC106041.1 - 1 - 2 296 71

View Full Product Details