Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IFNA5
Entrez Gene ID
3442
Gene Name
interferon alpha 5
Gene Aliases
IFN-alpha-5, IFN-alphaG, INA5, INFA5, leIF G
Chromosome Location
Chr.9: 21304614 - 21305313 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002169.2 NP_002160.1 1 - 1 453 68
GenBank mRNA BC093755.1 - 1 - 1 453 68
BC093757.1 - 1 - 1 453 68
V00541.1 - 1 - 1 228 68

View Full Product Details