The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001173984.2 | NP_001167455.1 | 9 - 10 | 1247 | 67 |
NM_013263.4 | NP_037395.2 | 9 - 10 | 1247 | 67 | |
XM_011523046.2 | XP_011521348.1 | 9 - 10 | 1333 | 67 | |
XM_011523047.2 | XP_011521349.1 | 9 - 10 | 1333 | 67 | |
XM_011523050.2 | XP_011521352.1 | 5 - 6 | 1353 | 67 | |
XM_017023181.1 | XP_016878670.1 | 5 - 6 | 1370 | 67 | |
GenBank mRNA | AF152604.1 | - | 9 - 10 | 1140 | 67 |
AF213969.1 | - | 9 - 10 | 1092 | 67 | |
AJ271881.1 | - | 9 - 10 | 1205 | 67 | |
AK027308.1 | - | 2 - 3 | 218 | 67 | |
AK074613.1 | - | 3 - 4 | 384 | 67 | |
BC001611.1 | - | 5 - 6 | 508 | 67 | |
BC050728.1 | - | 10 - 11 | 1154 | 67 | |
BC094706.1 | - | 10 - 11 | 1124 | 67 |