Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ASNS
Entrez Gene ID
440
Gene Name
asparagine synthetase (glutamine-hydrolyzing)
Gene Aliases
ASNSD, TS11
Chromosome Location
Chr.7: 97852117 - 97872542 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178075.1 NP_001171546.1 10 - 11 1422 102
NM_001178076.1 NP_001171547.1 8 - 9 1188 102
NM_001178077.1 NP_001171548.1 8 - 9 1121 102
NM_001673.4 NP_001664.3 10 - 11 1496 102
NM_133436.3 NP_597680.2 10 - 11 1751 102
NM_183356.3 NP_899199.2 11 - 12 1774 102
GenBank mRNA AK000379.1 - 10 - 11 1719 102
AK302189.1 - 10 - 11 1316 102
AK302242.1 - 8 - 9 1069 102
AK316224.1 - 8 - 9 1123 102
BC008723.2 - 11 - 12 1601 102
BC014621.2 - 10 - 11 1372 102
BT007113.1 - 8 - 9 1245 102
M15798.1 - 9 - 10 1292 102

View Full Product Details