Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SAMD7
Entrez Gene ID
344658
Gene Name
sterile alpha motif domain containing 7
Gene Aliases
-
Chromosome Location
Chr.3: 169911572 - 169939175 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001304366.1 NP_001291295.1 1 - 2 250 103
NM_182610.3 NP_872416.1 1 - 2 250 103
NR_130713.1 1 - 2 250 103
GenBank mRNA AL711948.1 - 1 - 2 251 103
BX537903.1 - 1 - 2 130 103
BX648534.1 - 1 - 2 131 103
BX648989.1 - 1 - 2 130 103
BX648991.1 - 1 - 2 251 103

View Full Product Details