The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001304366.1 | NP_001291295.1 | 7 - 8 | 1400 | 88 |
NM_182610.3 | NP_872416.1 | 7 - 8 | 1423 | 88 | |
NR_130713.1 | 8 - 9 | 1513 | 88 | ||
GenBank mRNA | BC117339.1 | - | 6 - 7 | 1107 | 88 |
BC143909.1 | - | 7 - 8 | 1219 | 88 | |
BX537903.1 | - | 7 - 8 | 1303 | 88 | |
BX648534.1 | - | 7 - 8 | 1281 | 88 | |
BX648989.1 | - | 7 - 8 | 1303 | 88 | |
BX648991.1 | - | 7 - 8 | 1401 | 88 |