Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- CADPS
- Entrez Gene ID
-
8618
- Gene Name
- calcium dependent secretion activator
- Gene Aliases
-
CADPS1, CAPS, CAPS1, UNC-31
- Chromosome Location
-
Chr.3: 62398346 - 62875389 on Build GRCh38
View Full Product Details