Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RMDN2
Entrez Gene ID
151393
Gene Name
regulator of microtubule dynamics 2
Gene Aliases
BLOCK18, FAM82A, FAM82A1, PRO34163, PYST9371, RMD-2, RMD2, RMD4
Chromosome Location
Chr.2: 37920789 - 38068032 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170791.2 NP_001164262.1 6 - 7 989 100
NM_001170792.2 NP_001164263.1 6 - 7 1060 100
NM_001170793.2 NP_001164264.1 5 - 6 566 100
NM_001322211.1 NP_001309140.1 6 - 7 1058 100
NM_001322212.1 NP_001309141.1 6 - 7 989 100
NM_144713.4 NP_653314.3 6 - 7 1535 100
XM_011532613.2 XP_011530915.1 6 - 7 1516 100
XM_011532614.2 XP_011530916.1 8 - 9 1647 100
XM_011532615.2 XP_011530917.2 8 - 9 1647 100
XM_011532616.2 XP_011530918.1 6 - 7 961 100
XM_011532617.2 XP_011530919.1 6 - 7 1047 100
XM_011532618.2 XP_011530920.1 7 - 8 1171 100
XM_011532619.2 XP_011530921.1 6 - 7 1054 100
XM_017003474.1 XP_016858963.1 8 - 9 1647 100
XM_017003475.1 XP_016858964.1 8 - 9 1647 100
XM_017003476.1 XP_016858965.1 6 - 7 961 100
XM_017003477.1 XP_016858966.1 6 - 7 1053 100
XM_017003478.1 XP_016858967.1 6 - 7 961 100
XM_017003479.1 XP_016858968.1 8 - 9 1140 100
GenBank mRNA AF435956.1 - 5 - 6 546 100
AK057516.1 - 6 - 7 1517 100
AK095462.1 - 7 - 8 1046 100
BC024243.2 - 6 - 7 1062 100

View Full Product Details