Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RMDN2
Entrez Gene ID
151393
Gene Name
regulator of microtubule dynamics 2
Gene Aliases
BLOCK18, FAM82A, FAM82A1, PRO34163, PYST9371, RMD-2, RMD2, RMD4
Chromosome Location
Chr.2: 37920789 - 38068032 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170791.2 NP_001164262.1 9 - 10 1225 97
NM_001170792.2 NP_001164263.1 9 - 10 1296 97
NM_001170793.2 NP_001164264.1 8 - 9 802 97
NM_001322211.1 NP_001309140.1 9 - 10 1294 97
NM_001322212.1 NP_001309141.1 9 - 10 1225 97
NM_144713.4 NP_653314.3 9 - 10 1771 97
XM_011532613.2 XP_011530915.1 9 - 10 1752 97
XM_011532614.2 XP_011530916.1 11 - 12 1883 97
XM_011532615.2 XP_011530917.2 11 - 12 1883 97
XM_011532616.2 XP_011530918.1 9 - 10 1197 97
XM_011532617.2 XP_011530919.1 9 - 10 1283 97
XM_011532618.2 XP_011530920.1 10 - 11 1407 97
XM_011532619.2 XP_011530921.1 9 - 10 1290 97
XM_017003474.1 XP_016858963.1 11 - 12 1883 97
XM_017003475.1 XP_016858964.1 11 - 12 1883 97
XM_017003476.1 XP_016858965.1 9 - 10 1197 97
XM_017003477.1 XP_016858966.1 9 - 10 1289 97
XM_017003478.1 XP_016858967.1 9 - 10 1197 97
XM_017003479.1 XP_016858968.1 11 - 12 1376 97
GenBank mRNA AF435956.1 - 8 - 9 782 97
AK057516.1 - 9 - 10 1753 97
AY358269.1 - 9 - 10 1225 97
BC024243.2 - 9 - 10 1298 97
DB523982.1 - 2 - 3 218 97

View Full Product Details